Air-O-Cell Cassette, Electron Microscopy Sciences
Air-O-Cell Cassette, Electron Microscopy Sciences
Certificates
Air-O-Cell Cassette, Electron Microscopy Sciences
Air-O-Cell Cassette, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This is a unique air sampling cassette specially designed for quick collection of a wide range of airborne aerosols including mold spores, pollen, insect parts, skin cell fragments, fibers (e.g., asbestos, fiberglass, cellulose, clothing fibers, and more.) and organic particulate e.g. ceramic, fly ash, copy toner, and more.
The Air-O-Cell® collects both viable and non-viable sample specimens, providing a much broader overview of potential allergens and contaminants than conventional sampling techniques. After sampling is completed, the cassettes are sent to a laboratory, where the slides are removed and direct microscopic analysis can be immediately performed.
The collection media is compatible with a wide range of biological stains and refractive index oils, allowing direct quantitative analysis of organic and inorganic particulate. The Air-O-Cell® can be used with any standard off-the-shelf area sampling pump capable of drawing 15 LPM open flow. The small compact size makes Air-O-Cell® suitable for use in confined or restrictive area.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Cassette
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.51
    $218.51
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $160.78
    $160.78
    Each
       $218.51
    Pack of 10
       $160.78
    Supplier Number: 65210-10
  • Cassette
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $831.19
    $831.19
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $592.96
    $592.96
    Each
       $831.19
    Pack of 50
       $592.96
    Supplier Number: 65210-50