This mounting medium is made with limonene, a natural product from orange peels. It preserves tissues and cell smears that can be dehydrated with organic solvents in immunohistochemistry, e.g. DAB and DAB with nickel or cobalt.
- A direct substitute for BioMeda™ Clarion Mount
- Use for mounting tissues and cell smears
- Works well with alkaline phosphatase chromogens, and organic solvent resistant Supper Fast Red
- A good choice for mounting H & E stained slides.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- UN No.:1993
- Storage temperature:2-8°C