Borosilicate Capillary Micro Glass Slide, Electron Microscopy Sciences
Borosilicate Capillary Micro Glass Slide, Electron Microscopy Sciences
Borosilicate Capillary Micro Glass Slide, Electron Microscopy Sciences
Certificates
Borosilicate Capillary Micro Glass Slide, Electron Microscopy Sciences
Borosilicate Capillary Micro Glass Slide, Electron Microscopy Sciences
11 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from Corning® 7740 borosilicate, low expansion, type I glass. Micro glass slides are precision rectangular glass capillary tubes for use in microscopic viewing of liquid samples.
  • packaged in plastic vials
  • Internal diameter tolerance is ±10%
  • Wall tolerance is ±20%
  • Withstand heat up to 500 °C
Capillary action instantly collects the sample and the thin glass walls minimize evaporation. The design of these cells eliminates the needs for cover glass and the hanging drop method. Samples can be retained for future reference by sealing or cementing open ends.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Épaisseur
    L×l
    Availability
    Price
  • Capy Boro slides, colored ends
    White
    0.02 mm
    50×0.2 mm
    Discontinued
    Supplier Number: 63820-05
  • Capy Boro slides, colored ends
    Blue
    0.02 mm
    100×0.2 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $203.78
    $203.78
    Log in pour la disponibilité
    Pack of 30
    Pack of 30 - $153.67
    $153.67
    Each
       $203.78
    Pack of 30
       $153.67
    Supplier Number: 63820-10
  • Capy Boro slides, colored ends
    Green
    0.02 mm
    50×0.3 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $135.88
    $135.88
    Log in pour la disponibilité
    Pack of 33
    Pack of 33 - $103.25
    $103.25
    Each
       $135.88
    Pack of 33
       $103.25
    Supplier Number: 63821-05
  • Capy Boro slides, colored ends
    Yellow
    0.03 mm
    100×0.3 mm
    Discontinued
    Supplier Number: 63821-10
  • Capy Boro slides, colored ends
    White
    0.03 mm
    50×0.4 mm
    Discontinued
    Supplier Number: 63822-05
  • Capy Boro slides, colored ends
    Blue
    0.04 mm
    100×0.4 mm
    Discontinued
    Supplier Number: 63822-10
  • Capy Boro slides, colored ends
    Green
    0.04 mm
    50×0.5 mm
    Discontinued
    Supplier Number: 63823-05
  • Capy Boro slides, colored ends
    Yellow
    0.05 mm
    100×0.5 mm
    Discontinued
    Supplier Number: 63823-10
  • Capy Boro slides, colored ends
    White
    0.05 mm
    50×1.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $162.24
    $162.24
    Log in pour la disponibilité
    Pack of 40
    Pack of 40 - $123.70
    $123.70
    Each
       $162.24
    Pack of 40
       $123.70
    Supplier Number: 63824-05
  • Capy Boro slides, colored ends
    Blue
    0.10 mm
    100×1.0 mm
    Discontinued
    Supplier Number: 63824-10
  • Capy Boro slides, colored ends
    Green
    0.10 mm
    50×2.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $154.64
    $154.64
    Log in pour la disponibilité
    Pack of 36
    Pack of 36 - $117.66
    $117.66
    Each
       $154.64
    Pack of 36
       $117.66
    Supplier Number: 63825-05
  • Capy Boro slides, colored ends
    Yellow
    0.20 mm
    100×2.0 mm
    Discontinued
    Supplier Number: 63825-10
  • Capy Boro slides, colored ends
    White
    0.20 mm
    50×3.0 mm
    Discontinued
    Supplier Number: 63826-05
  • Capy Boro slides, colored ends
    Blue
    0.30 mm
    100×3.0 mm
    Discontinued
    Supplier Number: 63826-10
  • Capy Boro slides, colored ends
    Green
    0.30 mm
    50×4.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $189.52
    $189.52
    Log in pour la disponibilité
    Pack of 28
    Pack of 28 - $144.70
    $144.70
    Each
       $189.52
    Pack of 28
       $144.70
    Supplier Number: 63827-05
  • Capy Boro slides, colored ends
    Yellow
    0.40 mm
    100×4.0 mm
    Discontinued
    Supplier Number: 63827-10
  • Capy Boro slides, colored ends
    White
    0.30 mm
    0.50×5.0 mm
    Discontinued
    Supplier Number: 63828-10
  • Capy Boro slides, colored ends
    Blue
    0.50 mm
    100×6.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $180.09
    $180.09
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $136.86
    $136.86
    Each
       $180.09
    Pack of 12
       $136.86
    Supplier Number: 63829-10
  • Capy Boro slides, colored ends
    Green
    0.60 mm
    100×7.0 mm
    Discontinued
    Supplier Number: 63830-10
  • Capy Boro slides, colored ends
    Yellow
    0.70 mm
    100×2.0 mm
    Discontinued
    Supplier Number: 63831-10
  • Capy Boro slides, colored ends
    White
    0.10 mm
    100×9.0 mm
    Discontinued
    Supplier Number: 63832-10
  • Capy Boro slides, colored ends
    Blue
    0.90 mm
    100×10.0 mm
    Discontinued
    Supplier Number: 63833-10
  • Capy Boro slides, colored ends
    Green
    1.00 mm
    50×1.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $176.14
    $176.14
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $141.66
    $141.66
    Each
       $176.14
    Pack of 50
       $141.66
    Supplier Number: 63840-05
  • Capy Boro slides, colored ends
    Yellow
    0.05 mm
    100×1.0 mm
    Discontinued
    Supplier Number: 63840-10
  • Capy Boro slides, colored ends
    White
    0.05 mm
    50×2.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $195.01
    $195.01
    Log in pour la disponibilité
    Pack of 45
    Pack of 45 - $148.88
    $148.88
    Each
       $195.01
    Pack of 45
       $148.88
    Supplier Number: 63841-05
  • Capy Boro slides, colored ends
    Blue
    0.10 mm
    100×2.0 mm
    Discontinued
    Supplier Number: 63841-10
  • Capy Boro slides, colored ends
    Green
    0.20 mm
    50×4.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $159.19
    $159.19
    Log in pour la disponibilité
    Pack of 36
    Pack of 36 - $121.36
    $121.36
    Each
       $159.19
    Pack of 36
       $121.36
    Supplier Number: 63842-05
  • Capy Boro slides, colored ends
    Yellow
    0.20 mm
    100×4.0 mm
    Discontinued
    Supplier Number: 63842-10
  • Capy Boro slides, colored ends
    White
    0.30 mm
    50×6.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $155.07
    $155.07
    Log in pour la disponibilité
    Pack of 24
    Pack of 24 - $117.98
    $117.98
    Each
       $155.07
    Pack of 24
       $117.98
    Supplier Number: 63843-05
  • Capy Boro slides, colored ends
    Blue
    0.30 mm
    100×6.0 mm
    Discontinued
    Supplier Number: 63843-10
  • Capy Boro slides, colored ends
    Green
    0.40 mm
    50×8.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $138.86
    $138.86
    Log in pour la disponibilité
    Pack of 18
    Pack of 18 - $105.65
    $105.65
    Each
       $138.86
    Pack of 18
       $105.65
    Supplier Number: 63844-05
  • Capy Boro slides, colored ends
    Yellow
    0.40 mm
    100×8.0 mm
    Discontinued
    Supplier Number: 63844-10