Vibration Dampening Mount, Electron Microscopy Sciences | Avantor
Vibration Dampening Mount, Electron Microscopy Sciences
Vibration Dampening Mount, Electron Microscopy Sciences
Certificates
Vibration Dampening Mount, Electron Microscopy Sciences
Vibration Dampening Mount, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Isolates sensitive analytical balances and other instruments from disturbing variations. Economical space saving alternative to full size balance tables.
  • Platform 64 mm (2.5") thick
  • Supports up to 16 kg (35 lbs.)
  • Overall height 76 mm (3")
  • Two sizes available
Polished surface resists scratches and chemical attack. Molded from dense black & white terrazzo. Four Vibro-Absorber cushions and neoprene feet.
Product Family Options
Informations sur le produit
  • Désignation
    W×D
    Availability
    Price
  • Vibration Damping Mount
    18×22"
    Discontinued
    Supplier Number: -
  • Vibration Damping Mount, 13¾ x 13¾"
    13¾×13¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,449.24
    $3,449.24
    Each
       $3,449.24
    Supplier Number: 67120-02