CoolSafe Deluxe Kits, Electron Microscopy Sciences | Avantor
CoolSafe Deluxe Kits, Electron Microscopy Sciences
CoolSafe Deluxe Kits, Electron Microscopy Sciences
Certificates
CoolSafe Deluxe Kits, Electron Microscopy Sciences
CoolSafe Deluxe Kits, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Each complete CoolSafe Deluxe system includes 1 CoolSafe Deluxe box, 1 Cool Chamber, and 2 Cool Bricks (1°C & –10°C).
  • Additional sample protection
  • Smaller bench-top footprint
  • Improved portability
  • Clear lid for sample visibility
CoolSafe for 0.2 mL PCR plates or tubes and the CoolSafe™ Deluxe box also include 4 holes for 1.5 mL tubes and 6 holes for 0.5 mL tubes.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • CoolSafe™ Kit for 2 ml tubes or PCR Plates
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,061.47
    $1,061.47
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - $1,061.47
    $1,061.47
    Each
       $1,061.47
    Kit of 2
       $1,061.47
    Supplier Number: 61954-10
  • CoolSafe™ Kit for 0.5 ml tubes
    Discontinued
    Supplier Number: 61954-20