Feather® Safeshield™ Scalpels, Electron Microscopy Sciences | Avantor
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Certificates
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from heat-treated stainless steel, these scalpels are excellent for delicate precise procedures
  • Heat-treated
  • Comfortable hand fit
  • Retractable shield
  • Color-coded blade size identification
They fit the hand comfortably and are designed with safety in mind. They reduce injury and transmitted disease from contaminated blades. They feature a clear protective, retractable shield and have a color coded button on the protective shield which assists with blade size identification.
Packaging: Individually packaged, 10 per box.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Feather® Safeshield™ Blade #10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $100.51
    $100.51
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $76.85
    $76.85
    Each
       $100.51
    Pack of 10
       $76.85
    Supplier Number: 72043-10
  • Feather® Safeshield™ Blade #11
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $100.51
    $100.51
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $76.85
    $76.85
    Each
       $100.51
    Pack of 10
       $76.85
    Supplier Number: 72043-11
  • Feather® Safeshield™ Blade #14
    Discontinued
    Supplier Number: 72043-14
  • Feather® Safesheild™ Blade #15
    Discontinued
    Supplier Number: 72043-15
  • Feather® Safeshield™ Blade #15c
    Discontinued
    Supplier Number: -
  • Feather® Safeshield™ Blade #20
    Discontinued
    Supplier Number: 72043-20
  • Feather® Safeshield™ Blade #21
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $113.73
    $113.73
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $86.98
    $86.98
    Each
       $113.73
    Pack of 10
       $86.98
    Supplier Number: 72043-21
  • Feather® Safeshield™ Blade #22
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $113.14
    $113.14
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $86.52
    $86.52
    Each
       $113.14
    Pack of 10
       $86.52
    Supplier Number: 72043-22
  • Feather® Safeshield™ Blade #25
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $112.23
    $112.23
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $85.83
    $85.83
    Each
       $112.23
    Pack of 10
       $85.83
    Supplier Number: 72043-25