Tri-Seal™ container, Electron Microscopy Sciences
About this item
Tri-Seal™ Specimen Container is a patented, leakproof, airtight, touch-top specimen container for the pathology lab.
Vials have permanent hinge and triple seal design. These are molded from 100% High-Density, translucent polypropylene and are available in a variety of sizes from 13 ml to 300 ml.
1 Options Available Below
Product Details & Documents
Tri-Seal™ Specimen Container is a patented, leakproof, airtight, touch-top specimen container for the pathology lab.
Vials have permanent hinge and triple seal design. These are molded from 100% High-Density, translucent polypropylene and are available in a variety of sizes from 13 ml to 300 ml.
Product Family Options
Product Information
- CapacitéDésignationDimensionsAvailabilityPrice
- 120 ml67 × 60 mmDiscontinued-Supplier Number: 64234-120LSpecifications:
More
Specifications
Cat. No.102094-974 - 15 ml21 × 60 mmEach$1,396.96Pack of 500$868.91Supplier Number: 64234-15Specifications:
More
Specifications
Cat. No.102094-976 - 15 mlBulkDiscontinued-Supplier Number: 64234-15BSpecifications:
More
Specifications
Cat. No.102094-978 - 240 ml76.2 × 76.2 mmDiscontinued-Supplier Number: 64234-240LSpecifications:
More
Specifications
Cat. No.102094-980 - 90 ml60 × 60 mmDiscontinued-Supplier Number: 64234-90LSpecifications:
More
Specifications
Cat. No.102094-982 - 120 ml63 × 50 mmDiscontinued-Supplier Number: 64235-120Specifications:
More
Specifications
Cat. No.102094-984 - 120 mlTall43 × 90 mmDiscontinued-Supplier Number: 64235-120TSpecifications:
More
Specifications
Cat. No.102094-986 - 120 mlTall With Locking Latch43 × 90 mmDiscontinued-Supplier Number: 64235-120TLSpecifications:
More
Specifications
Cat. No.102094-988 - 13 ml15 × 95 mmDiscontinued-Supplier Number: 64235-13Specifications:
More
Specifications
Cat. No.102094-990 - 240 ml75 × 75 mmDiscontinued-Supplier Number: 64235-240Specifications:
More
Specifications
Cat. No.102094-992 - 300 mlTall With Locking Latch30 × 80 mmDiscontinued-Supplier Number: 64235-300TLSpecifications:
More
Specifications
Cat. No.102094-994 - 45 mlTamper Evident Transport Container30 × 80 mmDiscontinued-Supplier Number: 64235-45TESpecifications:
More
Specifications
Cat. No.102094-996 - 50 mlTall28 × 84 mmDiscontinued-Supplier Number: 64235-50TSpecifications:
More
Specifications
Cat. No.102094-998 - 50 mlTall With Locking Latch28 × 84 mmDiscontinued-Supplier Number: 64235-50TLSpecifications:
More
Specifications
Cat. No.102095-000 - 75 mlTall33 × 93 mmDiscontinued-Supplier Number: 64235-75TSpecifications:
More
Specifications
Cat. No.102095-002 - 90 ml50 × 50 mmDiscontinued-Supplier Number: 64235-90Specifications:
More
Specifications
Cat. No.102095-004