Sealing Films, Electron Microscopy Sciences
Sealing Films, Electron Microscopy Sciences
Sealing Films, Electron Microscopy Sciences
Certificates
Sealing Films, Electron Microscopy Sciences
Sealing Films, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Use sealing films to eliminate well-to-well contamination, microbial and viral contamination, and sample spillage. Non breathable membranes prevent evaporation.
  • Sterilized by gamma irradiation and free of cytotoxins
  • Two scored end-tabsmake film positioning and removal hassle-free
  • Thermal Stability to withstand a wide temperature range
  • Pierceable sealing films facilitate easy removal of samples with a pipet tip
Breathe-Easy® is an adhesive, gas-permeable, cell culture membrane. It is made of 25.4 µm thick polyurethane. It is UV transparent to 300 nm and can be used in temperature range of -80 to 100 °C.
Breathe-EASIER™ is a rapid gas transmissable sealing membrane for cell growth. Breathe-Easier membranes permit rapid, consistent gas exchange of Oxygen, CO2 and water vapor for uniform cell. It is made of 139.7 µm thick non-woven rayon. It is not UV transparent and can be used in temperature range of -20 to 80 °C.
AluminaSeal™ is non-cytotoxic and sterile and is resistant to DMSO, chemicals, solvents and other common reagents. It can be used for Cold and PCR Storage in a temperature range of -80 to 110 °C. It is made of 68.6 µm aluminum and is not gas permeable or UV transparent.
HotSeal™ is a PCR sealing film for use in the temperature range of -40 to 125 °C. It is made of 58.4 µm polyester and is not gas permeable or UV transparent.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Plage de température
    Épaisseur
    L×l
    Availability
    Price
  • Breathe-Easy Cell Culture Membrane, for use with Cell Culture
    PU
    –80...+100 °C
    25.4 µm
    6×3.25"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $331.40
    $331.40
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $252.58
    $252.58
    Each
       $331.40
    Pack of 100
       $252.58
    Supplier Number: 70536-10
  • HotSeal Sealing Film, for use with PRC
    Polyester
    –40...+125 °C
    58.4 µm
    6×3.25"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $196.30
    $196.30
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $149.97
    $149.97
    Each
       $196.30
    Pack of 100
       $149.97
    Supplier Number: 70534-20
  • AluminaSeal Sealing Film, for use with Cold Storage
    Aluminum
    –80...+110 °C
    68.6 µm
    6×3.25"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $251.92
    $251.92
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $192.55
    $192.55
    Each
       $251.92
    Pack of 100
       $192.55
    Supplier Number: 70534-10
  • Breathe-EASIER Sealing Membrane for Cell Growth, for use with Cell Culture
    Non-woven rayon
    –20…+80 °C
    139.7 µm
    6×3.25"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $299.96
    $299.96
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $229.18
    $229.18
    Each
       $299.96
    Pack of 100
       $229.18
    Supplier Number: 70536-20