About this item
15 & 50 mL Chambers
Ideal for use on hot or cold plates or for temperature sensitive liquid transfers. Anodized aluminum blocks will not rust or corrode. A variety of options are available to accommodatePCR tubes and trays, 1.5 mL tubes, 15 mL and 50 mL conical tubes. A variety of Specialty Chambers are available to keep samples cool and on ice while you work. Freeze these Chambers overnight for added cooling convenience. These Chambers do not fit the CoolSafe system; use separately (*61955-30 not pictured).
15 & 50 mL Chambers
Each Chamber is an anodized aluminum block. Excellent temperature conductors. Use with hot plates or with our Ice Pans & Ice Buckets.
Double Chamber for 0.2 mL tubes
The Double Chamber fits two 96-well PCR trays or individual tubes and strips. Each edge can accommodate up to (12) 0.5 mL and (8) 1.5 mL tubes. Great for temperature sensitive liquid transfers. It measures 6 3/16 x 6 1/2 x 1".
64-Place Step Chamber
Description: Anodized aluminum block sized to fit 64 1.5 mL microtubes (16 holes per 'step') for easy viewing of samples when in the freezer, on the bench-top or in an ice pan or ice bucket.
80-Well Chamber
The 80-well Chamber is a solid, anodized aluminum block sized to fit 1.5 mL microtubes. An excellent temperature conductor for use with hot plates and ice buckets. It measures 8.25 x 2.625 x 1.25".
Double Chamber for 0.2 mL tubes
The Double Chamber fits two 96-well PCR trays or individual tubes and strips. Each edge can accommodate up to (12) 0.5 mL and (8) 1.5 mL tubes. Great for temperature sensitive liquid transfers. It measures 6 3/16 x 6 1/2 x 1".
64-Place Step Chamber
Description: Anodized aluminum block sized to fit 64 1.5 mL microtubes (16 holes per 'step') for easy viewing of samples when in the freezer, on the bench-top or in an ice pan or ice bucket.
80-Well Chamber
The 80-well Chamber is a solid, anodized aluminum block sized to fit 1.5 mL microtubes. An excellent temperature conductor for use with hot plates and ice buckets. It measures 8.25 x 2.625 x 1.25".
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.