Thermal Transfer Labels, Electron Microscopy Sciences
À propos de cet article
Thermal Transfer Cryo-Tags are a reliable and convenient way to identify samples with text or a barcode on microtubes, microplates, bottles and other laboratory containers. Pre-cut, peel-off labels designed for use in thermal transfer printers. Print popular barcode formats for fast and easy sample identification. Works with all thermal transfer printers.
- Unique solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance
- Specially coated for use with a resin ribbon that provides the best performance under extreme conditions
- Resists Degradation from scuffing, chemicals, moisture and wide temperature fluctuations
- Thermal Transfer Labels for Storage in Temperatures From −196°C to 80°C
- Print as little as one label at a time, eliminating label waste and saving time and money
- Print crisp, clear, professional quality labels
8 Options Available Below
Product Details & Documents
Thermal Transfer Cryo-Tags are a reliable and convenient way to identify samples with text or a barcode on microtubes, microplates, bottles and other laboratory containers. Pre-cut, peel-off labels designed for use in thermal transfer printers. Print popular barcode formats for fast and easy sample identification. Works with all thermal transfer printers.
- Unique solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance
- Specially coated for use with a resin ribbon that provides the best performance under extreme conditions
- Resists Degradation from scuffing, chemicals, moisture and wide temperature fluctuations
- Thermal Transfer Labels for Storage in Temperatures From −196°C to 80°C
- Print as little as one label at a time, eliminating label waste and saving time and money
- Print crisp, clear, professional quality labels
Product Family Options
Informations sur le produit
- DésignationPour l'utilisation avecDimensionsAvailabilityPrice
- Thermal transfer labels0.2 ml PCR tubes20.6 x 7 mmDiscontinued-Supplier Number: 77592-65
- Thermal transfer labels0.2 ml PCR tubes20.6 x 7 mmDiscontinued-Supplier Number: 77592-81
- Thermal transfer labels0.5 ml tubes24 x 13 mmDiscontinued-Supplier Number: 77592-70
- Thermal transfer labels0.5 ml tubes24 x 13 mmDiscontinued-Supplier Number: 77592-82
- Thermal transfer labels0.5 ml tubes27 x 13 mmEach$266.17Roll of 2,000$266.17Supplier Number: 77592-2
- Thermal transfer labels0.5 ml tubes27 x 13 mmDiscontinued-Supplier Number: 77592-84
- Thermal transfer labels1.5 - 2 ml tubes32 x 13 mmEach$368.32Roll of 2,000$368.32Supplier Number: 77592-75
- Thermal transfer labels1.5 - 2 ml tubes32 x 13 mmEach$600.20Roll of 4,000$600.20Supplier Number: 77592-83
- Thermal transfer labels1.5 - 2 ml tubes35 x 16 mmDiscontinued-Supplier Number: 77592-80
- Thermal transfer labels1.5 - 2 ml tubes35 x 16 mmEach$462.27Roll of 4,000$462.27Supplier Number: 77592-85
- Thermal transfer labels1.5 - 2 ml tubes38 x 13 mmEach$399.82Roll of 2,000$399.82Supplier Number: 77592-3
- Thermal transfer labels1.5 - 2 ml tubes38 x 13 mmDiscontinued-Supplier Number: 77592-86
- Thermal transfer labelsGeneral purpose38 x 19 mmEach$425.73Roll of 2,000$425.73Supplier Number: 77592-4
- Thermal transfer labelsGeneral purpose38 x 19 mmEach$871.17Roll of 4,000$871.17Supplier Number: 77592-87
- Thermal transfer labelsMicroplates51 x 6 mmEach$247.10Roll of 2,000$247.10Supplier Number: 77592-5
- Thermal transfer labelsMicroplates51 x 6 mmDiscontinued-Supplier Number: 77592-88
- Thermal transfer labelsDiscontinued-Supplier Number: 77592-1