SupaMega Cassette, Electron Microscopy Sciences
SupaMega Cassette, Electron Microscopy Sciences
SupaMega Cassette, Electron Microscopy Sciences
SupaMega Cassette, Electron Microscopy Sciences
Ideal for prostate, brain, breast tissue and eyes.
Extremely useful for research and toxicology laboratories.
Measures:
Inside: 64 mm x 48 mm x 13 mm
Outside: 75 mm x 52 mm x 18 mm
Product Family Options
  • Désignation
    Color
  • SupaMega Cassette
    Blue
    Each
    Each - $360.80
    $360.80
    Pack of 100
    Pack of 100 - $274.79
    $274.79
    Each
       $360.80
    Pack of 100
       $274.79
     : 70065-B
  • SupaMega Cassette
    Green
    Each
    Each - $369.64
    $369.64
    Pack of 100
    Pack of 100 - $281.51
    $281.51
    Each
       $369.64
    Pack of 100
       $281.51
     : 70065-G
  • SupaMega Cassette
    Pink
     : 70065-P
  • SupaMega Cassette
    White
    Each
    Each - $371.34
    $371.34
    Pack of 100
    Pack of 100 - $282.80
    $282.80
    Each
       $371.34
    Pack of 100
       $282.80
     : 70065-W
  • SupaMega Cassette
    Yellow
    Each
    Each - $374.94
    $374.94
    Pack of 100
    Pack of 100 - $285.54
    $285.54
    Each
       $374.94
    Pack of 100
       $285.54
     : 70065-Y