Automatic Pipette Washing Set, Electron Microscopy Sciences | Avantor
Automatic Pipette Washing Set, Electron Microscopy Sciences
Automatic Pipette Washing Set, Electron Microscopy Sciences
Certificates
Automatic Pipette Washing Set, Electron Microscopy Sciences
Automatic Pipette Washing Set, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Solve the problem of washing pipettes quickly, safely, and with little effort. To use allow dirty pipettes to soak in the pipette basket in the pipette jar and when ready the basket is transferred to the washer where a constant flow of tap water rinses the pipettes clean.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Pipette Basket
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $375.21
    $375.21
    Each
       $375.21
    Supplier Number: 64377-25
  • Pipette Jar
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $467.29
    $467.29
    Each
       $467.29
    Supplier Number: 64377-15
  • Pipette Washer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,668.76
    $4,668.76
    Each
       $4,668.76
    Supplier Number: 64377-05