Desiccators Canister, Electron Microscopy Sciences | Avantor
Desiccators Canister, Electron Microscopy Sciences
Silica Gel Canister, 14×18 mm
Catalog # 100489-242
Fournisseur #:  71205-01
Desiccators Canister, Electron Microscopy Sciences
Catalog # 100489-242
Desiccators Canister, Electron Microscopy Sciences
Desiccators Canister, Electron Microscopy Sciences
Silica Gel Canister, 14×18 mm
Price
$63.68
Log in pour la disponibilité
Each Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This small container offers more desiccant in less space. It is ship-proof and possesses greater drying capabilities. Designed for use in bottles and small containers. Perfect for protecting against potency loss, odor formation, color changes, caking, mildew and condensation.
  • Provides superior absorption
  • Remains dry at maximum saturation
  • Silica gel can absorb 15% of its weight in water vapor in two hours
  • Silica Gel-Desiccant is a fast and extra moisture absorbing agent
Capsule shell is made from polyethylene with very fine mesh top and bottom. The canister contains 1 gram silica gel.
Spécifications
  • Désignation:
    Silica Gel Canister
  • Diameter ×H:
    14×18 mm