Tungsten Boat, Electron Microscopy Sciences
Tungsten Boat, Electron Microscopy Sciences
Tungsten Boat, Electron Microscopy Sciences
Certificates
Tungsten Boat, Electron Microscopy Sciences
Tungsten Boat, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tungsten has the highest melting point and lowest vapor pressure of all metals, and at the temperatures over 1650°C has the highest tensile strength
Tungsten is the most useful material used in vacuum evaporation technique, due to its special properties and cost factor.
It has excellent corrosion resistance and is attacked only slightly by most mineral acids.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Overall Length
    Épaisseur
    Availability
    Price
  • Tungsten Boat B-2
    12.7L × 9.5W × 3.2D (1⁄2 × 5⁄8 × 1⁄32)"
    101.6×19.05 mm (4×¾")
    0.25 mm (0.01")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $256.58
    $256.58
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $200.85
    $200.85
    Each
       $256.58
    Pack of 5
       $200.85
    Supplier Number: 73816-B2
  • Tungsten Boat B-5
    9.65 × 3.81 mm
    101.6×12.7 mm (4×½")
    0.25 mm (0.01")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $195.94
    $195.94
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $147.59
    $147.59
    Each
       $195.94
    Pack of 5
       $147.59
    Supplier Number: 73816-B5
  • Tungsten Boat
    12.7L × 9.5W × 3.2D mm
    32 mm (1¼")
    0.05 mm (0.002")
    Discontinued
    Supplier Number: 73816
  • Tungsten Boat B-3
    39.1L × 6.35W × 3.3H mm
    101.6×19.05 mm (4×¾")
    0.25 mm (0.01")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $237.14
    $237.14
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $180.93
    $180.93
    Each
       $237.14
    Pack of 5
       $180.93
    Supplier Number: 73816-B3
  • Tungsten Boat B-4
    28.9L × 15.87W × 3.3D
    100.5×19.05 mm (4×¾")
    0.25 mm (0.01")
    Discontinued
    Supplier Number: 73816-B4
  • Tungsten Boat B-6
    39.1L × 12W × 3.55D mm
    101.6×39.1 mm (4×½")
    0.25 mm (0.01")
    Discontinued
    Supplier Number: 73816-B6
  • Tungsten Boat B-7
    53.84L × 25W × 2.28D mm
    101.6×25.4 mm (4×1")
    0.25 mm (0.01")
    Discontinued
    Supplier Number: 73816-B7