Tray Set with Plastic Boxes, Electron Microscopy Sciences
À propos de cet article
A compact, solid fiberboard tray box makes instituting a plastic box/sample storage system more convenient and efficient.
- Available with or without accompanying plastic boxes
To form a tray set, use plastic boxes for specimen storage, and then organize boxes within the fiberboard tray Box. These sets eliminates misplacement or inadvertent sample loss and allows for rapid retrieval of specimens.
7 Options Available Below
Product Details & Documents
A compact, solid fiberboard tray box makes instituting a plastic box/sample storage system more convenient and efficient.
- Available with or without accompanying plastic boxes
To form a tray set, use plastic boxes for specimen storage, and then organize boxes within the fiberboard tray Box. These sets eliminates misplacement or inadvertent sample loss and allows for rapid retrieval of specimens.
Product Family Options
Informations sur le produit
- DésignationTailleAvailabilityPrice
- Supplier Number: 64350-02
- Supplier Number: 64350-09
- Supplier Number: 64350-03
- Tray Set With Plastic Boxes20 Box SetDiscontinued-Supplier Number: 64350-07
- Tray Set100 Box/SetDiscontinued-Supplier Number: 64350-01
- Supplier Number: 64350-04
- Supplier Number: 64350-06
- Supplier Number: 64350-08
- Supplier Number: 64350-05