Partitioned Plastic Boxes, Electron Microscopy Sciences
Partitioned Plastic Boxes, Electron Microscopy Sciences
Partitioned Plastic Boxes, Electron Microscopy Sciences
Partitioned Plastic Boxes, Electron Microscopy Sciences
Partitioned Plastic Boxes, Electron Microscopy Sciences
Organize singular or groups of items in individual compartments.
  • Extra-thick wall construction permits containment of heavy objects, protects delicate items
Stored items stay separately and are readily visible at all times.
Product Family Options
  • Désignation
    Dimensions
  • 18 Compartments
    11×6-3/4×1-3/4" (Overall); 1⅝×1-15/16×1-9/16" (Compartments)
    Each
    Each - $93.36
    $93.36
    Each
       $93.36
     : 64343
  • 12 Compartments
    7×3-3/4×1-1/4" (Overall); 1-11/16×1-1/16×1-1/8" (Compartments)
    Each
    Each - $48.10
    $48.10
    Each
       $48.10
     : 64341
  • 8 Compartments
    4-5/8×3×1-1/8" (Overall); "1-1/16×1-1/4×1"" (Compartments)
     : -
  • 36 Compartments
    10×7×1-13/16" (Overall);1½×1-1/16×1-9/16" (Compartments)
    Each
    Each - $113.93
    $113.93
    Each
       $113.93
     : 64344
  • 5 Compartments
    5-7/8×3-1/2×15⁄16" (OVerall); 1-1/16×3×1-1/8" (Compartments)
    Each
    Each - $50.88
    $50.88
    Each
       $50.88
     : 64340
  • 24 Compartments
    13-1/8×9×2-5/16" (Overall); "2×2×2-1/16" (Compartments)
    Each
    Each - $156.32
    $156.32
    Each
       $156.32
     : 64346
  • 4 Compartments
    4-5/8×3×1-1/8" (Overall);1-5/16×2-1/8×1" (Compartments)
    Each
    Each - $43.20
    $43.20
    Each
       $43.20
     : 64339-04
  • 18 Compartments
    8-1/4×4 ½×1 3⁄8" (Overall); 1-1/4×1-1/4×1-3/16" (Compartments)
    Each
    Each - $63.19
    $63.19
    Each
       $63.19
     : 64342
  • 12 Compartments
    13-1/8×9×2-5/16" (Overall); 2×4-3/16×2-1/16" (Compartments)
    Each
    Each - $138.59
    $138.59
    Each
       $138.59
     : 64345
  • 6 Compartments
    4-5/8×3×1-1/8" (Overall); "1-5/16×1-3/8×1"" (Compartments)
    Each
    Each - $46.11
    $46.11
    Each
       $46.11
     : 64339-06