Serum Bottles, Electron Microscopy Sciences
À propos de cet article
Clear bottles are manufactured from '400' borosilicate molded glass. Amber bottles are manufactured from '500' borosilicate molded glass.
Certifications: Meets USP Type I standards.
8 Options Available Below
Product Details & Documents
Clear bottles are manufactured from '400' borosilicate molded glass. Amber bottles are manufactured from '500' borosilicate molded glass.
Certifications: Meets USP Type I standards.
Product Family Options
Informations sur le produit
- CapacitéDésignationColorAvailabilityPrice
- 20 mlSerum BottlesAmberCase of 288$1,723.85Each$1,723.85Supplier Number: 60308-20
- 5 mlSerum BottlesClearCase of 288$763.12Each$763.12Supplier Number: 60306-05
- 30 mlSerum BottlesClearCase of 288$1,617.06Each$1,617.06Supplier Number: 60306-30
- 100 mlSerum BottlesAmberCase of 144$2,274.51Each$2,274.51Supplier Number: 60308-100
- 30 mlSerum BottlesAmberCase of 288$2,071.89Each$2,071.89Supplier Number: 60308-30
- 20 mlSerum BottlesClearDiscontinued-Supplier Number: 60306-20
- 5 mlSerum BottlesAmberCase of 288$1,550.41Each$1,550.41Supplier Number: 60308-05
- 10 mlSerum BottlesAmberCase of 288$1,725.91Each$1,725.91Supplier Number: 60308-10
- 10 mlSerum BottlesClearDiscontinued-Supplier Number: 60306-10
- 50 mlSerum BottlesClearDiscontinued-Supplier Number: 60306-50
- 2 mlSerum BottlesClearCase of 288$664.26Each$664.26Supplier Number: 60306-02