Pinholes, Electron Microscopy Sciences | Avantor
Pinholes, Electron Microscopy Sciences
Pinholes, Electron Microscopy Sciences
Certificates
Pinholes, Electron Microscopy Sciences
Pinholes, Electron Microscopy Sciences
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These pinholes are prepared from pure copper foil, 3 mm in diameter, and 25 microns thick.
  • Possess very high roundness and edge retention
  • Blackened on one surface
  • Mounted in black anodized aluminum discs
These grids are used in spatial filtering, controlling the diameter of light beams, creating point light sources and Image analysis.
Mounted in a recessed hole in an anodized holder, these pin holes measure 16 mm in diameter.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Pinhole, 5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $743.91
    $743.91
    Each
       $743.91
    Supplier Number: PH-C5
  • Pinhole, 10 µm
    Discontinued
    Supplier Number: PH-10
  • Pinhole, 25 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $566.64
    $566.64
    Each
       $566.64
    Supplier Number: PH-C25
  • Pinhole, 100 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $517.62
    $517.62
    Each
       $517.62
    Supplier Number: PH-100
  • Pinhole, 250 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $477.19
    $477.19
    Each
       $477.19
    Supplier Number: PH-C250
  • Pinhole, 1000 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $477.19
    $477.19
    Each
       $477.19
    Supplier Number: PH-C1000