Master Coiler Plier, Electron Microscopy Sciences
Master Coiler Plier, Electron Microscopy Sciences
Master Coiler Plier, Stainless steel jaws, 5.75"
Catalog # 100498-002
Supplier #:  62186-20
Master Coiler Plier, Electron Microscopy Sciences
Catalog # 100498-002
Master Coiler Plier, Electron Microscopy Sciences
Master Coiler Plier, Electron Microscopy Sciences
Master Coiler Plier, Stainless steel jaws, 5.75"
Price
$34.17
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This ʽMaster Coiler’ is the painless plier. It is designed to make loops and coils without nicking precious wire.
  • Comfort grips
  • Non-rust
It allows one to hold the wire with the flat jaw while rotating the round jaw to form the perfect loop.
Specifications
  • Désignation:
    Master Coiler Plier, Stainless steel jaws
  • Longueur:
    5.75"