Multipurpose Electron Microscope Specimen Box – MEM Grid Box, Electron Microscopy Sciences | Avantor
Multipurpose Electron Microscope Specimen Box – MEM Grid Box, Electron Microscopy Sciences
Multipurpose Electron Microscope Specimen Box – MEM Grid Box, Electron Microscopy Sciences
Certificates
Multipurpose Electron Microscope Specimen Box – MEM Grid Box, Electron Microscopy Sciences
Multipurpose Electron Microscope Specimen Box – MEM Grid Box, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A newly designed Grid Box with safety, ease and convenience in mind. This multipurpose electron microscope specimen grid box is one of the most desirable boxes on the market.
Eliminates the chances of tweezer insertion damaging the grids – The "tweezers slot" and "grid slot" are in a separate location. The tweezers are only able to grip the edge of the grid enabling it to be picked up.
Grids stored no longer jump out of the box while you remove the cover – Between the body of the box and the sliding lid, there is a separate plastic cover, which allows for only four slots being exposed at a time.
The Grid Record Card is stored safely by insertion along on the reverse side of the box
MEM-96 – will able to store up to 96 grids
MEM-32 – will able to store up to 32 grids with 8 Blocks
Measures: 81 mm (L) x 54 mm (W) x 6 mm Thick
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • MEM-96 Grid Storage Box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.35
    $35.35
    Each
       $35.35
    Supplier Number: 71164-01
  • MEM-96 Grid Storage Box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $416.68
    $416.68
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $357.79
    $357.79
    Each
       $416.68
    Pack of 10
       $357.79
    Supplier Number: 71164-10
  • MEM-32 Grid Storage Box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $42.57
    $42.57
    Each
       $42.57
    Supplier Number: 71165-01