Nunc™ 384 Well® Optical Bottom Plates – Polymer Base, Electron Microscopy Sciences | Avantor
Nunc™ 384 Well® Optical Bottom Plates – Polymer Base, Electron Microscopy Sciences
Nunc™ 384 Well® Optical Bottom Plates – Polymer Base, Electron Microscopy Sciences
Certificates
Nunc™ 384 Well® Optical Bottom Plates – Polymer Base, Electron Microscopy Sciences
Nunc™ 384 Well® Optical Bottom Plates – Polymer Base, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The NUNC™ Coverglass Optical Bottom 96- and 384- Well Plates have an optically clear polystyrene plate bottom
This clear base permits microscopic viewing and reading of well The plate’s upper structure is offered in black (for fluorometric detection).
Product Family Options
Informations sur le produit
  • Désignation
    Working Well Volume
    Dimensions
    Color
    Surface Treatment
    Availability
    Price
  • Non-Sterile
    10 - 100 µl
    128 × 86 mm
    Black
    Untreated
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - 1 634,69 $
    1 634,69 $
    Log in pour la disponibilité
    Each
    Each - 1 634,69 $
    1 634,69 $
    Case of 30
       1 634,69 $
    Each
       1 634,69 $
    Supplier Number: 64811-20
  • Sterile with Lid
    10 - 100 µl
    128 × 86 mm
    Black
    Cell culture
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - 1 490,20 $
    1 490,20 $
    Log in pour la disponibilité
    Each
    Each - 1 490,20 $
    1 490,20 $
    Case of 30
       1 490,20 $
    Each
       1 490,20 $
    Supplier Number: 64810-35
  • Non-Sterile with lid
    10 - 100 µl
    128 × 86 mm
    White
    Collagen I
    Discontinued
    Supplier Number: -
  • Non-Sterile with lid
    10 - 100 µl
    128 × 86 mm
    Black
    Collagen I
    Discontinued
    Supplier Number: 64810-52
  • Non-Sterile with lid
    10 - 100 µl
    128 × 86 mm
    Black
    Poly-D-Lysine
    Discontinued
    Supplier Number: 64810-54