Zeiss SEM Mounts, Electron Microscopy Sciences
Zeiss SEM Mounts, Electron Microscopy Sciences
Zeiss SEM Mounts, Electron Microscopy Sciences
Certificates
Zeiss SEM Mounts, Electron Microscopy Sciences
Zeiss SEM Mounts, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Flat End Pin Surface
  • Diameter: 12.7 mm (1/2")
  • Slotted Head diameter, 3.1 mm (1/8") Pin
Product Family Options
Product Information
  • Désignation
    Matière
    Availability
    Price
  • Zeiss Flat End Pin
    Aluminum
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - 102,24 $
    102,24 $
    Log in for availability
    Pack of 50
    Pack of 50 - 83,41 $
    83,41 $
    Each
       102,24 $
    Pack of 50
       83,41 $
    Supplier Number: 75500
  • Zeiss Flat End Pin
    Aluminum
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - 160,93 $
    160,93 $
    Log in for availability
    Pack of 100
    Pack of 100 - 134,47 $
    134,47 $
    Each
       160,93 $
    Pack of 100
       134,47 $
    Supplier Number: 75510
  • Zeiss Flat End Pin
    Aluminum
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - 669,27 $
    669,27 $
    Log in for availability
    Pack of 500
    Pack of 500 - 560,14 $
    560,14 $
    Each
       669,27 $
    Pack of 500
       560,14 $
    Supplier Number: 75520