Hitachi S-500 Threaded Pin SEM Mounts, Electron Microscopy Sciences
Product Details & Documents
Threaded pin with the head dia. of 25 mm (1") and pin dia. of 6 mm (1/4").
Specifications
- Désignation:Hitachi S-500
- Matière:Aluminum