AMRAY 1400 SEM Mounts, Electron Microscopy Sciences
À propos de cet article
Slotted Head 12.7 mm (1/2") diameter. Pin 3.1 mm (1/8") diameter.
3 Options Available Below
Product Details & Documents
Slotted Head 12.7 mm (1/2") diameter. Pin 3.1 mm (1/8") diameter.
Product Family Options
Informations sur le produit
- DésignationMatièreAvailabilityPrice
- Amray 1400AluminumEach$139.66Pack of 50$117.66Supplier Number: 75172
- Amray 1400AluminumEach$236.17Pack of 100$198.38Supplier Number: 75174
- Amray 1400CarbonEach$78.18Pack of 10$72.05Supplier Number: 76170