Lyophilization Seals, Electron Microscopy Sciences
À propos de cet article
The Lyophilization seals come in four styles.
The Center Disc Tear Out style exposes the top of the stopper to the syringe. Natural in color. The Flip-Cap, Center Tear style is easy-to-use. Push upward on plastic button to access stopper or septum. No sharp metal edges. Removal of the Flip Cap Button can be accomplished with one hand, freeing the other for the syringe. The Complete Tear Off style seals can be completely removed and contents poured from the bottle. The Lined Aluminum Seals are ideal for filling or retrieving contents with a syringe and as closures for many instrumentation applications. These seals have a center disc tear-out, seal lined with "PTFE" – Faced natural red rubber.
3 Options Available Below
Product Details & Documents
The Lyophilization seals come in four styles.
The Center Disc Tear Out style exposes the top of the stopper to the syringe. Natural in color. The Flip-Cap, Center Tear style is easy-to-use. Push upward on plastic button to access stopper or septum. No sharp metal edges. Removal of the Flip Cap Button can be accomplished with one hand, freeing the other for the syringe. The Complete Tear Off style seals can be completely removed and contents poured from the bottle. The Lined Aluminum Seals are ideal for filling or retrieving contents with a syringe and as closures for many instrumentation applications. These seals have a center disc tear-out, seal lined with "PTFE" – Faced natural red rubber.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Unlined Aluminum Seals, 7x13Discontinued-Supplier Number: 60320-07
- Lined Aluminum Seals, 13 NaturalEach$225.34Pack of 100$161.70Supplier Number: 60330-13N
- Flipcap Center Tear, 7x13Discontinued-Supplier Number: 60321-07
- Complete Tear Off, 13x20Each$37.44Pack of 100$26.92Supplier Number: 60322-13
- Complete Tear Off, 7x13Each$37.44Pack of 100$26.92Supplier Number: 60322-07