Disposable Pipettes – Sterile, Electron Microscopy Sciences
À propos de cet article
Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to ±1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
5 Options Available Below
Product Details & Documents
Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to ±1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
Product Family Options
Informations sur le produit
- VolumePackedAvailabilityPrice
- 1 mlSingle WrapEach$133.14Pack of 100$100.82Supplier Number: 70953-1
- 5 mlSingle WrapEach$96.49Pack of 50$73.52Supplier Number: 70953-5
- 2 mlBulkDiscontinued-Supplier Number: 70954-2
- 10 mlSingle WrapEach$91.32Pack of 50$68.59Supplier Number: 70953-10
- 10 mlBulkEach$68.43Pack of 25$52.11Supplier Number: 70954-10
- 50 mlSingle WrapEach$510.63Pack of 50$385.84Supplier Number: 70953-50
- 1 mlBulkDiscontinued-Supplier Number: -