Extra Long Transfer Pipettes, Electron Microscopy Sciences | Avantor
Extra Long Transfer Pipettes, Electron Microscopy Sciences
Extra Long Transfer Pipettes, Electron Microscopy Sciences
Certificates
Extra Long Transfer Pipettes, Electron Microscopy Sciences
Extra Long Transfer Pipettes, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These transfer pipettes are ideal for subculturing, harvesting, sampling 15, 50, 100 ml tubes and flasks, removing supernatant and media from cell cultures, wash cells, post centrifugation to remove cell layers, layer or transfers in columns, loading gels, field testing, and blood culture bottle. These pipettes come sterile and non-sterile. Sterile pipettes are individually wrapped , then are bulk packaged.
Product Family Options
Informations sur le produit
  • Volume
    Ø×L
    Sterility
    Drop Size
    Bulb Draw
    Availability
    Price
  • 23.0 ml
    8.6×30.0 cm
    Non sterile
    20 µL
    7.3 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $109.45
    $109.45
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $78.49
    $78.49
    Each
       $109.45
    Pack of 100
       $78.49
    Supplier Number: 70968-27
  • 6.0 ml
    5.0×22.5 cm
    Sterile
    25 µL
    2.3 ml
    Discontinued
    Supplier Number: 70968-26S