General Purpose Transfer Pipette, Electron Microscopy Sciences
General Purpose Transfer Pipette, Electron Microscopy Sciences
Certificates
General Purpose Transfer Pipette, Electron Microscopy Sciences
General Purpose Transfer Pipette, Electron Microscopy Sciences
11 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These pipettes are good for blood banking, reagent handling, viscous liquid sampling and transferring, etc. The non-sterile pipettes are bulk packed and the sterile are individually packed.
Product Family Options
Product Information
  • Volume
    Ø×L
    Sterility
    Bulb Draw
    Availability
    Price
  • 1.7 ml
    4.8×8.7 cm
    Non sterile
    0.9 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $83.94
    $83.94
    Log in for availability
    Pack of 500
    Pack of 500 - $60.02
    $60.02
    Each
       $83.94
    Pack of 500
       $60.02
    Supplier Number: 70960-3
  • 9.3 ml
    6.1×13.7 cm
    Non sterile
    4.8 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $61.67
    $61.67
    Log in for availability
    Pack of 250
    Pack of 250 - $43.21
    $43.21
    Each
       $61.67
    Pack of 250
       $43.21
    Supplier Number: 70960-6
  • 8.0 ml
    6.1×15.7 cm
    Non sterile
    4.6 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $94.36
    $94.36
    Log in for availability
    Pack of 400
    Pack of 400 - $70.78
    $70.78
    Each
       $94.36
    Pack of 400
       $70.78
    Supplier Number: 70960-2
  • 4.0 ml
    6.6×12.9 cm
    Non sterile
    1.9 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $96.17
    $96.17
    Log in for availability
    Pack of 400
    Pack of 400 - $68.78
    $68.78
    Each
       $96.17
    Pack of 400
       $68.78
    Supplier Number: 70960-4
  • 4.0 ml
    6.6×15.4 cm
    Non sterile
    1.9 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $97.59
    $97.59
    Log in for availability
    Pack of 500
    Pack of 500 - $69.79
    $69.79
    Each
       $97.59
    Pack of 500
       $69.79
    Supplier Number: 70960-5
  • 7.7 ml
    7.9×15.6 cm
    Non sterile
    3.2 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $102.55
    $102.55
    Log in for availability
    Pack of 500
    Pack of 500 - $77.04
    $77.04
    Each
       $102.55
    Pack of 500
       $77.04
    Supplier Number: 70960-1
  • 1.7 ml
    4.8×8.7 cm
    Sterile
    0.9 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $217.33
    $217.33
    Log in for availability
    Pack of 500
    Pack of 500 - $158.88
    $158.88
    Each
       $217.33
    Pack of 500
       $158.88
    Supplier Number: 70960-3S
  • 9.3 ml
    6.1×13.7 cm
    Sterile
    4.8 ml
    Discontinued
    Supplier Number: 70960-6S
  • 8.0 ml
    6.1×15.7 cm
    Sterile
    4.6 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $213.83
    $213.83
    Log in for availability
    Pack of 100
    Pack of 100 - $150.97
    $150.97
    Each
       $213.83
    Pack of 100
       $150.97
    Supplier Number: 70960-2S
  • 4.0 ml
    6.6×12.9 cm
    Sterile
    1.9 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $211.48
    $211.48
    Log in for availability
    Pack of 400
    Pack of 400 - $156.21
    $156.21
    Each
       $211.48
    Pack of 400
       $156.21
    Supplier Number: 70960-4S
  • 4.0 ml
    6.6×15.4 cm
    Sterile
    1.9 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $202.80
    $202.80
    Log in for availability
    Pack of 500
    Pack of 500 - $150.24
    $150.24
    Each
       $202.80
    Pack of 500
       $150.24
    Supplier Number: 70960-5S
  • 7.7 ml
    7.9×15.6 cm
    Sterile
    3.2 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $197.36
    $197.36
    Log in for availability
    Pack of 500
    Pack of 500 - $149.31
    $149.31
    Each
       $197.36
    Pack of 500
       $149.31
    Supplier Number: 70960-1S