Thermotech™ PTFE Beakers, Electron Microscopy Sciences | Avantor
Thermotech™ PTFE Beakers, Electron Microscopy Sciences
Thermotech™ PTFE Beakers, Electron Microscopy Sciences
Certificates
Thermotech™ PTFE Beakers, Electron Microscopy Sciences
Thermotech™ PTFE Beakers, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This product features a unique combination of a pure "PTFE" body with a specially formulated stabilized "PTFE" carbon base as a single molding.
  • A totally inert pure PTFE inner surface
  • Heatable to 270°C without distortion
  • Improved heat transfer
All "PTFE" beaker caps can be used for Thermotech™ "PTFE" beakers, accordingly to its sizes.
Product Family Options
Informations sur le produit
  • Capacité
    Hauteur
    Ø ext.
    Availability
    Price
  • 400 ml
    112 mm
    85 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $377.79
    $377.79
    Each
       $377.79
    Supplier Number: 60930-40
  • 250 ml
    94 mm
    75 mm
    Discontinued
    Supplier Number: 60930-25