Stirring Rods, Electron Microscopy Sciences | Avantor
Stirring Rods, Electron Microscopy Sciences
Stirring Rods, Electron Microscopy Sciences
Certificates
Stirring Rods, Electron Microscopy Sciences
Stirring Rods, Electron Microscopy Sciences
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These stirring rods are available in two versions: pure 'PTFE' solid rod with tapered end and pure 'PTFE' with bendable steel core.
The version with steel core can be bent into a permanent shape. Rods are inert and will not scratch, use up to 280 °C.
Product Family Options
Informations sur le produit
  • Désignation
    Ø
    Longueur
    Availability
    Price
  • PTFE with Steel Core
    6 mm
    100 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $33.38
    $33.38
    Each
       $33.38
    Supplier Number: 66200-10
  • Solid PTFE Rod
    8 mm
    100 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.42
    $39.42
    Each
       $39.42
    Supplier Number: 66201-10
  • PTFE with Steel Core
    6 mm
    150 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $45.05
    $45.05
    Each
       $45.05
    Supplier Number: 66200-15
  • PTFE with Steel Core
    6 mm
    200 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $45.25
    $45.25
    Each
       $45.25
    Supplier Number: 66200-20
  • PTFE with Steel Core
    6 mm
    300 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $63.10
    $63.10
    Each
       $63.10
    Supplier Number: 66200-30
  • PTFE with Steel Core
    6 mm
    400 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $87.08
    $87.08
    Each
       $87.08
    Supplier Number: 66200-40