SuperFrost™ Clipped Corner Slide, Electron Microscopy Sciences
About this item
Designed specially for the Sysmex SP-100 slide prep unit and recommended for the Diagnostics.
The 45 degree clipped corner slide offers exceptional cleanliness and resistance to sticking. The tight tolerances for the length and width help to reduce accumulation of glass chips within the instrument. The pre-cleaned slide is 25×75 mm and approximately 1 mm thick. Ideal for technicians, who routinely prepare large number of slides.
2 Options Available Below
Product Details & Documents
Designed specially for the Sysmex SP-100 slide prep unit and recommended for the Diagnostics.
The 45 degree clipped corner slide offers exceptional cleanliness and resistance to sticking. The tight tolerances for the length and width help to reduce accumulation of glass chips within the instrument. The pre-cleaned slide is 25×75 mm and approximately 1 mm thick. Ideal for technicians, who routinely prepare large number of slides.
Product Family Options
Product Information
- DésignationCapacitéAvailabilityPrice
- Superfrost clipped corner1 gCase of 1,440$1,414.91Each$1,414.91Supplier Number: 71882-10
- Superfrost clipped corner1 gEach$165.38Pack of 144$124.77Supplier Number: 71882-01