Slide Rack, Electron Microscopy Sciences
Slide Rack, Electron Microscopy Sciences
Marketsource
Slide Rack, Electron Microscopy Sciences
Marketsource
Certificates
Slide Rack, Electron Microscopy Sciences
Slide Rack, Electron Microscopy Sciences
10 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Poly-ceramic slide rack designed for histology,cytology staining and coverslipping.
  • Made from Poly-Ceramic
  • Microwave safely in a distortion-free rack
  • Chemical resistant to all solvents below 200 °C
  • Designed – for histology and cytology staining and coverslipping
  • Tested for performance criteria and dimensional accuracy
  • Fit the Tissue-Tek Tape Machines
  • Easy slide insertion in V-Groove slots
Stainless Steel and Plastic Slide Racks are very good chemically as well as thermally but have limited uses in microwave and solvents. When it comes to inserting the slide, easy to break the glass or tear the tape on abrupt edges.
WINLAB Slide Racks are made from Poly-ceramic that maintains its structural integrity to 200 °C in any solvent while being microwaved. The racks last a lot longer than any other materials. WINLAB Racks save time and money.
Product Family Options
Product Information
  • Désignation
    Capacité
    Availability
    Price
  • Rack Carrier Blue
    20 Slides
    Log in for availability
    Log in for availability
    Each
    Each - $122.03
    $122.03
    Each
       $122.03
    Supplier Number: 71435-B
  • Wire Handle
    20 Slides
    Log in for availability
    Log in for availability
    Each
    Each - $70.93
    $70.93
    Each
       $70.93
    Supplier Number: 71437-20
  • Winlab Rack, LSM-20
    20 Slides
    Log in for availability
    Log in for availability
    Each
    Each - $162.38
    $162.38
    Each
       $162.38
    Supplier Number: 71431-20
  • Rack Carrier, Blue
    30 slides
    Log in for availability
    Log in for availability
    Each
    Each - $180.22
    $180.22
    Each
       $180.22
    Supplier Number: 71433-B
  • Winlab Rack, LS-20
    20 Slides
    Log in for availability
    Log in for availability
    Each
    Each - $152.73
    $152.73
    Each
       $152.73
    Supplier Number: 71430-20
  • Winlab Rack, LS-30
    30 slides
    Log in for availability
    Log in for availability
    Each
    Each - $216.00
    $216.00
    Each
       $216.00
    Supplier Number: 71430-30
  • SR-20 Plastic Handle
    20 Slides
    Log in for availability
    Log in for availability
    Each
    Each - $52.97
    $52.97
    Each
       $52.97
    Supplier Number: 71437-40
  • SR30 Plastic handle 30 slide unit
    30 slides
    Log in for availability
    Log in for availability
    Each
    Each - $59.94
    $59.94
    Each
       $59.94
    Supplier Number: 71437-50
  • Rack Carrier
    30 slides
    Log in for availability
    Log in for availability
    Each
    Each - $160.29
    $160.29
    Each
       $160.29
    Supplier Number: 71433-DAKO
    Specifications:
    Marketsource
    Marketsource
  • Rack Carrier
    20 Slides
    Log in for availability
    Log in for availability
    Each
    Each - $155.87
    $155.87
    Each
       $155.87
    Supplier Number: 71435-DAKO
    Specifications:
    Marketsource
    Marketsource