Ear Punch, Electron Microscopy Sciences
Catalogue # 101413-586
Fournisseur: Electron Microscopy Sciences
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Ear Punch
- Cat. no.:101413-586
- Supplier No.:72893-01
Spécifications
À propos de cet article
Made from stainless steel. Allows for you to make a 2 mm hole through the animal ear in just one second. It also can be used as a specimen puncher.