Cryo Labels on Sheets, Electron Microscopy Sciences
Cryo Labels on Sheets, Electron Microscopy Sciences
Cryo Labels on Sheets, Electron Microscopy Sciences
Certificates
Cryo Labels on Sheets, Electron Microscopy Sciences
Cryo Labels on Sheets, Electron Microscopy Sciences
29 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Cryo-Tag® labels are tough and temperature resistant withstanding freezer temperatures down to –196 °C (liquid and vapor phase nitrogen), boiling water baths (100 °C), autoclaving and moderate oven temperatures (150 °C). Made with a flexible, polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations resulting in a strong adhesive bond.
  • Chemically inert labels resist most organic solvents and caustic agents
  • Adhere to most plastics*, glass, and metals without cracking, peeling or degrading
  • Label sheets are specially top-coated to readily accept laser printing
  • Heat-resistant, stay-flat sheets will not jam in laser printers
  • Temperature-Resistant : From −196 to 150 °C
*100% Virgin Polypropylene tubes are recommended. Avoid the use of siliconized surfaces.
Ordering information: Certificates of analysis are available.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Pour l'utilisation avec
    Dimensions
    Packaging
    Availability
    Price
  • Cryo labels, on sheets
    White
    Microscope slides
    25 x 25 mm
    80 labels/sheet, 20 sheets, (1600 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $215.32
    $215.32
    Log in pour la disponibilité
    Pack of 1,600
    Pack of 1,600 - $175.84
    $175.84
    Each
       $215.32
    Pack of 1,600
       $175.84
    Supplier Number: 77571-W
  • Cryo labels, on sheets
    White
    1.5 tubes
    33 x 13 mm with 9.5 mm spot
    68 labels/sheet, 20 sheets (1360 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $210.25
    $210.25
    Log in pour la disponibilité
    Pack of 1,360
    Pack of 1,360 - $178.38
    $178.38
    Each
       $210.25
    Pack of 1,360
       $178.38
    Supplier Number: 77574-W
  • Cryo labels, on sheets
    Rainbow
    Microplates
    38 x 6 mm
    156 labels/sheet, 20 sheets (3120 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.42
    $218.42
    Log in pour la disponibilité
    Pack of 3,120
    Pack of 3,120 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,120
       $178.38
    Supplier Number: 77567-05
  • Cryo labels, on sheets
    White
    Microplates
    38 x 6 mm
    156 labels/sheet, 20 sheets (3120 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $217.44
    $217.44
    Log in pour la disponibilité
    Pack of 3,120
    Pack of 3,120 - $177.56
    $177.56
    Each
       $217.44
    Pack of 3,120
       $177.56
    Supplier Number: 77567-W
  • Cryo labels, on sheets
    Yellow
    Microplates
    38 x 6 mm
    156 labels/sheet, 20 sheets (3120 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $239.75
    $239.75
    Log in pour la disponibilité
    Pack of 3,120
    Pack of 3,120 - $271.63
    $271.63
    Each
       $239.75
    Pack of 3,120
       $271.63
    Supplier Number: 77567-Y
  • Cryo labels, on sheets
    Rainbow
    General purpose
    38 x 19 mm
    60 labels/sheet, 20 sheets (1200 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $196.63
    $196.63
    Log in pour la disponibilité
    Pack of 1,200
    Pack of 1,200 - $178.38
    $178.38
    Each
       $196.63
    Pack of 1,200
       $178.38
    Supplier Number: 77564-05
  • Cryo labels, on sheets
    Blue
    General purpose
    38 x 19 mm
    60 labels/sheet, 20 sheets (1200 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.42
    $218.42
    Log in pour la disponibilité
    Pack of 1,200
    Pack of 1,200 - $178.38
    $178.38
    Each
       $218.42
    Pack of 1,200
       $178.38
    Supplier Number: 77564-B
  • Cryo labels, on sheets
    Green
    General purpose
    38 x 19 mm
    60 labels/sheet, 20 sheets (1200 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $195.72
    $195.72
    Log in pour la disponibilité
    Pack of 1,200
    Pack of 1,200 - $177.56
    $177.56
    Each
       $195.72
    Pack of 1,200
       $177.56
    Supplier Number: 77564-G
  • Cryo labels, on sheets
    Orange
    General purpose
    38 x 19 mm
    60 labels/sheet, 20 sheets (1200 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.42
    $218.42
    Log in pour la disponibilité
    Pack of 1,200
    Pack of 1,200 - $178.38
    $178.38
    Each
       $218.42
    Pack of 1,200
       $178.38
    Supplier Number: 77564-O
  • Cryo labels, on sheets
    Red
    General purpose
    38 x 19 mm
    60 labels/sheet, 20 sheets (1200 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.42
    $218.42
    Log in pour la disponibilité
    Pack of 1,200
    Pack of 1,200 - $178.38
    $178.38
    Each
       $218.42
    Pack of 1,200
       $178.38
    Supplier Number: 77564-R
  • Cryo labels, on sheets
    White
    General purpose
    38 x 19 mm
    60 labels/sheet, 20 sheets (1200 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $216.40
    $216.40
    Log in pour la disponibilité
    Pack of 1,200
    Pack of 1,200 - $176.71
    $176.71
    Each
       $216.40
    Pack of 1,200
       $176.71
    Supplier Number: 77564-W
  • Cryo labels, on sheets
    Yellow
    General purpose
    38 x 19 mm
    60 labels/sheet, 20 sheets (1200 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.42
    $218.42
    Log in pour la disponibilité
    Pack of 1,200
    Pack of 1,200 - $178.38
    $178.38
    Each
       $218.42
    Pack of 1,200
       $178.38
    Supplier Number: 77564-Y
  • Cryo labels, on sheets
    Rainbow
    Cryo vials
    43 x 19 mm
    52 labels/sheet, 20 sheets (1040 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.42
    $218.42
    Log in pour la disponibilité
    Pack of 1,040
    Pack of 1,040 - $178.38
    $178.38
    Each
       $218.42
    Pack of 1,040
       $178.38
    Supplier Number: 77565-05
  • Cryo labels, on sheets
    Blue
    Cryo vials
    43 x 19 mm
    52 labels/sheet, 20 sheets (1040 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $217.44
    $217.44
    Log in pour la disponibilité
    Pack of 1,040
    Pack of 1,040 - $177.56
    $177.56
    Each
       $217.44
    Pack of 1,040
       $177.56
    Supplier Number: 77565-B
  • Cryo labels, on sheets
    Green
    Cryo vials
    43 x 19 mm
    52 labels/sheet, 20 sheets (1040 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $215.38
    $215.38
    Log in pour la disponibilité
    Pack of 1,040
    Pack of 1,040 - $175.89
    $175.89
    Each
       $215.38
    Pack of 1,040
       $175.89
    Supplier Number: 77565-G
  • Cryo labels, on sheets
    Orange
    Cryo vials
    43 x 19 mm
    52 labels/sheet, 20 sheets (1040 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $215.38
    $215.38
    Log in pour la disponibilité
    Pack of 1,040
    Pack of 1,040 - $175.89
    $175.89
    Each
       $215.38
    Pack of 1,040
       $175.89
    Supplier Number: 77565-O
  • Cryo labels, on sheets
    Red
    Cryo vials
    43 x 19 mm
    52 labels/sheet, 20 sheets (1040 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $215.38
    $215.38
    Log in pour la disponibilité
    Pack of 1,040
    Pack of 1,040 - $175.89
    $175.89
    Each
       $215.38
    Pack of 1,040
       $175.89
    Supplier Number: 77565-R
  • Cryo labels, on sheets
    White
    Cryo vials
    43 x 19 mm
    52 labels/sheet, 20 sheets (1040 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $216.40
    $216.40
    Log in pour la disponibilité
    Pack of 1,040
    Pack of 1,040 - $176.71
    $176.71
    Each
       $216.40
    Pack of 1,040
       $176.71
    Supplier Number: 77565-W
  • Cryo labels, on sheets
    Yellow
    Cryo vials
    43 x 19 mm
    52 labels/sheet, 20 sheets (1040 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.42
    $218.42
    Log in pour la disponibilité
    Pack of 1,040
    Pack of 1,040 - $178.38
    $178.38
    Each
       $218.42
    Pack of 1,040
       $178.38
    Supplier Number: 77565-Y
  • Cryo labels, on sheets
    White
    0.5 - 2 ml tubes
    64 x 13 mm
    60 labels/sheet, 20 sheets (1200 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.42
    $218.42
    Log in pour la disponibilité
    Pack of 1,200
    Pack of 1,200 - $178.38
    $178.38
    Each
       $218.42
    Pack of 1,200
       $178.38
    Supplier Number: 77572-W
  • Cryo labels, on sheets
    Rainbow
    Racks and boxes
    67 x 25 mm
    30 labels/sheet, 20 sheets (600 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $217.44
    $217.44
    Log in pour la disponibilité
    Pack of 600
    Pack of 600 - $177.56
    $177.56
    Each
       $217.44
    Pack of 600
       $177.56
    Supplier Number: 77566-05
  • Cryo labels, on sheets
    Blue
    Racks and boxes
    67 x 25 mm
    30 labels/sheet, 20 sheets (600 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.42
    $218.42
    Log in pour la disponibilité
    Pack of 600
    Pack of 600 - $178.38
    $178.38
    Each
       $218.42
    Pack of 600
       $178.38
    Supplier Number: 77566-B
  • Cryo labels, on sheets
    Green
    Racks and boxes
    67 x 25 mm
    30 labels/sheet, 20 sheets (600 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $220.77
    $220.77
    Log in pour la disponibilité
    Pack of 600
    Pack of 600 - $178.38
    $178.38
    Each
       $220.77
    Pack of 600
       $178.38
    Supplier Number: 77566-G
  • Cryo labels, on sheets
    Orange
    Racks and boxes
    67 x 25 mm
    30 labels/sheet, 20 sheets (600 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.42
    $218.42
    Log in pour la disponibilité
    Pack of 600
    Pack of 600 - $178.38
    $178.38
    Each
       $218.42
    Pack of 600
       $178.38
    Supplier Number: 77566-O
  • Cryo labels, on sheets
    Red
    Racks and boxes
    67 x 25 mm
    30 labels/sheet, 20 sheets (600 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $191.14
    $191.14
    Log in pour la disponibilité
    Pack of 600
    Pack of 600 - $156.09
    $156.09
    Each
       $191.14
    Pack of 600
       $156.09
    Supplier Number: 77566-R
  • Cryo labels, on sheets
    White
    Racks and boxes
    67 x 25 mm
    30 labels/sheet, 20 sheets (600 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $209.16
    $209.16
    Log in pour la disponibilité
    Pack of 600
    Pack of 600 - $175.84
    $175.84
    Each
       $209.16
    Pack of 600
       $175.84
    Supplier Number: 77566-W
  • Cryo labels, on sheets
    Yellow
    Racks and boxes
    67 x 25 mm
    30 labels/sheet, 20 sheets (600 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $216.97
    $216.97
    Log in pour la disponibilité
    Pack of 600
    Pack of 600 - $177.18
    $177.18
    Each
       $216.97
    Pack of 600
       $177.18
    Supplier Number: 77566-Y
  • Cryo labels, on sheets
    White
    Bottles and beakers
    76 x 51 mm
    12 labels/sheet, 20 sheets (240 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.42
    $218.42
    Log in pour la disponibilité
    Pack of 240
    Pack of 240 - $178.38
    $178.38
    Each
       $218.42
    Pack of 240
       $178.38
    Supplier Number: 77568-W
  • Cryo labels, on sheets
    White
    Blank sheet
    216 x 279 mm
    1 label/sheet, 20 sheets (20 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $214.89
    $214.89
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $175.14
    $175.14
    Each
       $214.89
    Pack of 20
       $175.14
    Supplier Number: 77570-W