Cryo Labels, Laser Printable, on Sheets, Electron Microscopy Sciences
À propos de cet article
Specially designed innovative labels for cryogenic storage. Made with a flexible, polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations resulting in a strong adhesive bond.
- Withstands freezer temperatures to –196 °C (liquid and vapor phase nitrogen), boiling water baths (100 °C), autoclaving and moderate oven temperatures (150 °C)
- Resist most organic solvents and caustic agents
- Adhere to plastics, glass and metals without cracking, peeling or degrading
- Specially top-coated to accept laser printing and other markings
- Heat-resistant, stay-flat sheets will not jam in laser printers
Virgin Polypropylene tubes are recommended. Avoid the use of siliconized surfaces.
Packaging: Rainbow packs with 4 sheets of each color: Blue, Green, Red, Yellow.
15 Options Available Below
Product Details & Documents
Specially designed innovative labels for cryogenic storage. Made with a flexible, polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations resulting in a strong adhesive bond.
- Withstands freezer temperatures to –196 °C (liquid and vapor phase nitrogen), boiling water baths (100 °C), autoclaving and moderate oven temperatures (150 °C)
- Resist most organic solvents and caustic agents
- Adhere to plastics, glass and metals without cracking, peeling or degrading
- Specially top-coated to accept laser printing and other markings
- Heat-resistant, stay-flat sheets will not jam in laser printers
Virgin Polypropylene tubes are recommended. Avoid the use of siliconized surfaces.
Packaging: Rainbow packs with 4 sheets of each color: Blue, Green, Red, Yellow.
Product Family Options
Informations sur le produit
- DésignationColorPour l'utilisation avecDimensionsAvailabilityPrice
- Cryo labels, on sheetsRainbow0.5 ml tubes24 x 13 mmEach$212.20Pack of 2,380$178.38Supplier Number: 77561-05
- Cryo labels, on sheetsBlue0.5 ml tubes24 x 13 mmEach$215.32Pack of 2,380$175.84Supplier Number: 77561-B
- Cryo labels, on sheetsGreen0.5 ml tubes24 x 13 mmEach$218.42Pack of 2,380$178.38Supplier Number: 77561-G
- Cryo labels, on sheetsOrange0.5 ml tubes24 x 13 mmEach$218.42Pack of 2,380$178.38Supplier Number: 77561-O
- Cryo labels, on sheetsRed0.5 ml tubes24 x 13 mmEach$210.25Pack of 2,380$178.38Supplier Number: 77561-R
- Cryo labels, on sheetsWhite0.5 ml tubes24 x 13 mmEach$209.27Pack of 2,380$177.56Supplier Number: 77561-W
- Cryo labels, on sheetsYellow0.5 ml tubes24 x 13 mmEach$195.30Pack of 2,380$177.18Supplier Number: 77561-Y
- Cryo labels, on sheetsRainbow1.5 ml tubes33 x 13 mmEach$229.10Pack of 1,700$176.71Supplier Number: 77560-05
- Cryo labels, on sheetsBlue1.5 ml tubes33 x 13 mmEach$210.25Pack of 1,700$178.38Supplier Number: 77560-B
- Cryo labels, on sheetsGreen1.5 ml tubes33 x 13 mmEach$217.44Pack of 1,700$177.56Supplier Number: 77560-G
- Cryo labels, on sheetsOrange1.5 ml tubes33 x 13 mmEach$196.63Pack of 1,700$178.38Supplier Number: 77560-O
- Cryo labels, on sheetsRed1.5 ml tubes33 x 13 mmEach$196.63Pack of 1,700$178.38Supplier Number: 77560-R
- Cryo labels, on sheetsWhite1.5 ml tubes33 x 13 mmEach$171.10Pack of 1,020$140.10Supplier Number: 77559-W
- Cryo labels, on sheetsWhite1.5 ml tubes33 x 13 mmEach$230.23Pack of 1,700$177.61Supplier Number: 77560-W
- Cryo labels, on sheetsYellow1.5 ml tubes33 x 13 mmEach$195.30Pack of 1,700$177.18Supplier Number: 77560-Y