Cryo Labels, Laser Printable, Electron Microscopy Sciences | Avantor
Cryo Labels, Laser Printable, Electron Microscopy Sciences
Cryo Labels, Laser Printable, Electron Microscopy Sciences
Certificates
Cryo Labels, Laser Printable, Electron Microscopy Sciences
Cryo Labels, Laser Printable, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Clear durable cryogenic storage labels allow the contents of vials beneath labels to remain visible.
  • Temperature-resistant –196 to 70 °C
  • Remains fully adhered down to –196 °C (liquid and vapor phase nitrogen)
  • Resist most organic solvents and caustic agents
  • Adhere to plastics, glass and metals without cracking, peeling or degrading
  • pecially top-coated to accept laser printing and other markings
  • Heat-resistant, stay-flat sheets will not jam in laser printers
Made with a flexible, vinyl label material and permanent acrylic adhesive. This unique combination expands and contracts together as temperatures fluctuate resulting in a strong and sustained adhesive bond.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Dimensions
    Packaging
    Availability
    Price
  • Cryo labels for laser printing
    Vinyl
    Clear
    0.5 ml tubes
    24 x 13 mm
    119 labels/sheet
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $238.67
    $238.67
    Log in pour la disponibilité
    Pack of 2,380
    Pack of 2,380 - $194.89
    $194.89
    Each
       $238.67
    Pack of 2,380
       $194.89
    Supplier Number: 77576-C
  • Cryo labels for laser printing
    Vinyl
    Clear
    1.5 ml tubes
    39 x 13 mm
    85 labels/sheet
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $236.41
    $236.41
    Log in pour la disponibilité
    Pack of 1,700
    Pack of 1,700 - $193.03
    $193.03
    Each
       $236.41
    Pack of 1,700
       $193.03
    Supplier Number: 77575-C