Gel-Pak® Storage/Carrier Box, Electron Microscopy Sciences
Gel-Pak® Storage/Carrier Box, Electron Microscopy Sciences
Certificates
Gel-Pak® Storage/Carrier Box, Electron Microscopy Sciences
Gel-Pak® Storage/Carrier Box, Electron Microscopy Sciences
8 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Gel-Box AD Series Carriers owe their unique device-holding capabilities to a proprietary GEL coating applied directly to the bottom of a hinged box. Devices are held once contact is made with the GEL surface and will stay in place, even if the carrier is tilted or jarred.
  • No need for custom-molded containers for each device size
  • Ideal for handling subassemblies, hybrid models and packaged devices
  • Available in Transparent, Conductive and Anti-Static carriers in multiple sizes
  • Retention Level available: x0, x1, x2, x3, x4, x5, and x8 (the higher the number the more retention capability)
The carriers are designed for manual off-loading with tweezers or by hand. GEL-PAK AD series are ideal for handling devices where no direct contact with the top of the device is important.The GEL-BOX Carriers may also be used for bare die handling, such as GaAs, PETs, laser diodes, and beam-lead devices where the die size is small (less than 1 mm) and tweezers or other specialized techniques are used for off-handling.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Anti-Static Hinged Box
    2×2×1/4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 136,95 $
    1 136,95 $
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - 954,65 $
    954,65 $
    Each
       1 136,95 $
    Pack of 50
       954,65 $
    Supplier Number: 64903-X4
  • Antistatic Gel-Box
    2×2×1/4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 139,73 $
    1 139,73 $
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - 1 015,45 $
    1 015,45 $
    Each
       1 139,73 $
    Pack of 50
       1 015,45 $
    Supplier Number: 64901-X3
  • Conductive Hinged Box
    2×2×1/4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 418,29 $
    1 418,29 $
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - 1 070,15 $
    1 070,15 $
    Each
       1 418,29 $
    Pack of 50
       1 070,15 $
    Supplier Number: 64901-X4
  • Conductive Hinged Box
    2×3×3/8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 026,64 $
    1 026,64 $
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - 783,60 $
    783,60 $
    Each
       1 026,64 $
    Pack of 50
       783,60 $
    Supplier Number: 64904-X4
  • Conductive Hinged Box
    6×4×1/2"
    Discontinued
    Supplier Number: 64910-X3
  • Conductive Hinged Box
    7×5×1"
    Discontinued
    Supplier Number: -
  • Conductive Hinged Box
    7 3/4×3 1/2×1"
    Discontinued
    Supplier Number: 64914-X3
  • Conductive Hinged Box with Anti-Static Top
    2×2×1/4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 258,92 $
    1 258,92 $
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - 950,12 $
    950,12 $
    Each
       1 258,92 $
    Pack of 50
       950,12 $
    Supplier Number: 64902-X4
  • Conductive Hinged Box with Anti-Static Top
    4.5×3.5×7/8"
    Discontinued
    Supplier Number: 64908-X3
  • Conductive Hinged Box with Anti-Static Top
    7×5×1"
    Discontinued
    Supplier Number: 64912-X3
  • Transparent Gel-Box
    3.5×4.5×7/8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 749,54 $
    1 749,54 $
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - 1 319,81 $
    1 319,81 $
    Each
       1 749,54 $
    Pack of 50
       1 319,81 $
    Supplier Number: 64907-X4
  • Transparent Gel-Box
    6×4×1/2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 716,95 $
    1 716,95 $
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - 1 309,03 $
    1 309,03 $
    Each
       1 716,95 $
    Pack of 50
       1 309,03 $
    Supplier Number: 64911-X3
  • Transparent Gel-Box
    10×3 1/2×5/8"
    Discontinued
    Supplier Number: 64915-X3
  • Transparent Hinged Box
    1×1×1/4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 840,38 $
    840,38 $
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - 747,55 $
    747,55 $
    Each
       840,38 $
    Pack of 50
       747,55 $
    Supplier Number: 64900-X4