Low Temperature Dessicator, Electron Microscopy Sciences
À propos de cet article
Dry, dust-free storage for use in refrigerators, freezers or at room temperature. These durable cabinets are made from 6.35 mm (1/4") acrylic with full-length hinges and stainless steel twist locking latches.
- Easy viewing clear units or dark bonze for light sensitive materials
- Deep, slide-in trays with perforated bottom
- Available in three sizes
5 Options Available Below
Product Details & Documents
Dry, dust-free storage for use in refrigerators, freezers or at room temperature. These durable cabinets are made from 6.35 mm (1/4") acrylic with full-length hinges and stainless steel twist locking latches.
- Easy viewing clear units or dark bonze for light sensitive materials
- Deep, slide-in trays with perforated bottom
- Available in three sizes
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 71222
- Low Temperature Desiccator Cabinet, BronzeDiscontinued-Supplier Number: 71220-A
- Supplier Number: 71220
- Supplier Number: 71222-A
- Supplier Number: 71221
- Supplier Number: 71221-A