4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
À propos de cet article
Biopsy processing cassettes with four square compartments each measuring 13 mm.
- Cover and base have about 1676 square openings maximizing fluid exchange and ensuring proper drainage
Each case contains four dispenser boxes of 250 cassettes with covers.
4 Options Available Below
Product Details & Documents
Biopsy processing cassettes with four square compartments each measuring 13 mm.
- Cover and base have about 1676 square openings maximizing fluid exchange and ensuring proper drainage
Each case contains four dispenser boxes of 250 cassettes with covers.
Product Family Options
Informations sur le produit
- ColorAvailabilityPrice
- OrangeDiscontinued-Supplier Number: 70075-O
- PinkEach$813.48Pack of 1,000$614.19Supplier Number: 70075-P
- PeachEach$813.48Pack of 1,000$614.19Supplier Number: 70075-PE
- TanEach$813.48Pack of 1,000$614.19Supplier Number: 70075-T
- Fluo-GreenDiscontinued-Supplier Number: -
- WhiteEach$800.43Pack of 1,000$718.49Supplier Number: 70075-W
- GreenDiscontinued-Supplier Number: 70075-G
- LilacDiscontinued-Supplier Number: 70075-L
- Fluo-PinkDiscontinued-Supplier Number: 70075-FP