Cryo Transport Box, Electron Microscopy Sciences | Avantor
Cryo Transport Box, Electron Microscopy Sciences
Cryo Transport Box, Electron Microscopy Sciences
Certificates
Cryo Transport Box, Electron Microscopy Sciences
Cryo Transport Box, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Cryo vial transport box is made from polystyrene and is ideal for transporting cryo vials.
  • Organizes small amounts of cryo vials
  • Durable construction
  • Holds 1.0 to 2.0 mL cryo vials
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Cryo Transport Box for 5 Tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $234.87
    $234.87
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $194.28
    $194.28
    Each
       $234.87
    Pack of 20
       $194.28
    Supplier Number: 62004-05
  • Cryo Transport Box for 10 Tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $163.96
    $163.96
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $122.89
    $122.89
    Each
       $163.96
    Pack of 10
       $122.89
    Supplier Number: 62004-10