Microsurgical Knife, Electron Microscopy Sciences | Avantor
Microsurgical Knife, Electron Microscopy Sciences
Microsurgical Knife, Electron Microscopy Sciences
Certificates
Microsurgical Knife, Electron Microscopy Sciences
Microsurgical Knife, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from tool steel wire. Sharpened to microsurgical quality.
The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.
Packaging: 6 blister packs to a box.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Micro Surgical Knife 30°
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $184.43
    $184.43
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $138.69
    $138.69
    Each
       $184.43
    Pack of 6
       $138.69
    Supplier Number: 72047-30
  • Micro Surgical Knife 45°
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $154.54
    $154.54
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $138.69
    $138.69
    Each
       $154.54
    Pack of 6
       $138.69
    Supplier Number: 72047-45
  • Micro Surgical Knife 15°
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $186.14
    $186.14
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $139.99
    $139.99
    Each
       $186.14
    Pack of 6
       $139.99
    Supplier Number: 72047-15