Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Lab Jacks
- Cat. no.:100498-354
- Supplier No.:63225-24
This LabJack provides stable height adjustment and support for items in the lab such as flasks, baths or hot plate stirrers.
Top and bottom plates are constructed of anodized aluminum while the support mechanism is made of chrome plated steel.