Hi Temperature Indicator Tape, Electron Microscopy Sciences
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Specialty cloth-textured Hi Temperature Indicator tape shows the word “STERILIZED” in bold, black letters after standard sterilization processing. The tape sticks to almost any surface or shape, holds fast in 550°F dry oven temperatures. Available in widths of 1/2", 3/4", and 1" on 250” rolls.
Product Family Options
  • Désignation
  • Hi-Temp Indicator Tape,½" Wide
    Each
    Each - $182.58
    $182.58
    Roll of 1
    Roll of 1 - $182.58
    $182.58
    Each
       $182.58
    Roll of 1
       $182.58
     : 77015-05
  • Hi-Temp Indicator Tape, ¾" Wide
    Each
    Each - $211.55
    $211.55
    Roll of 1
    Roll of 1 - $211.55
    $211.55
    Each
       $211.55
    Roll of 1
       $211.55
     : 77015-07
  • Hi-Temp Indicator Tape, 1" Wide
    Each
    Each - $215.83
    $215.83
    Roll of 1
    Roll of 1 - $215.83
    $215.83
    Each
       $215.83
    Roll of 1
       $215.83
     : 77015-10