Etched Double Neubauer, Electron Microscopy Sciences
Etched Double Neubauer, Electron Microscopy Sciences
Etched Double Neubauer, Electron Microscopy Sciences
Etched Double Neubauer, Electron Microscopy Sciences
Etched Double Neubauer, Electron Microscopy Sciences
Double improved Neubauer rulings are diamond etched directly into a polished recess to reduce damage from scratches. Opaque surfaces above and below the counting area offer an easy focal reference. The chamber comes with a quick orientation mark for the microscope. 0.1 mm cell depth.
 : The complete set includes one counting chamber, two 0.5 mm cover glasses, pipettes, and carrying case.
Product Family Options
  • Désignation
  • Chamber With Two Cover Glasses
    Each
    Each - $1,266.81
    $1,266.81
    Each
       $1,266.81
     : 63511-11
  • Chamber Only
    Each
    Each - $1,196.34
    $1,196.34
    Each
       $1,196.34
     : 63511-13
  • Ruled Hemacytometer, Complete Set
    Each
    Each - $1,821.36
    $1,821.36
    Each
       $1,821.36
     : 63511-10