Zerostat Anti-Static Gun, Electron Microscopy Sciences
Zero-Stat Anti-Static Gun
Supplier: Electron Microscopy Science
Fournisseur #: 60610
Zerostat Anti-Static Gun, Electron Microscopy Sciences
Catalog # 100496-120
Supplier: Electron Microscopy Science
Jump to:
Product Details & Documents
The Zerostat Anti-Static Gun operates on a piezoelectric effect to emit positive and negative charges to neutralize static on any surface. This anti-static gun emits no radioactivity and does not require electricity or batteries.
Measures: 4×6"
Documents
Spécifications
- Désignation:Zero-Stat Anti-Static Gun