Full Protractor, Electron Microscopy Sciences | Avantor
Full Protractor, Electron Microscopy Sciences
Full Protractor, Electron Microscopy Sciences
Certificates
Full Protractor, Electron Microscopy Sciences
Full Protractor, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
NE45 eyepiece scale is surface chrome picture, 10 mm diameter separated in degrees, 16 mm full protractor scale.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • NE45 Eyepiece Scale
    16 mm
    Discontinued
    Supplier Number: 68002-16
  • NE45 Eyepiece Scale
    19 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $377.80
    $377.80
    Each
       $377.80
    Supplier Number: 68002-19
  • NE45 Eyepiece Scale
    21 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $357.90
    $357.90
    Each
       $357.90
    Supplier Number: 68002-21