CryoVial®, Electron Microscopy Sciences
À propos de cet article
These vials are designed for storage of biological material, human or animal cells, at temperatures as low as –190 °C.
- The cap features a long skirt for one hand aseptic technique
- A super fast thread design that allows tightening or removal with a mere 1¼" turn, and an inside thread design that will not contribute to possible contamination
- Tubes are provided with white marking area for sample identification and can be color-coded by the use of a CAPINSERT®
- Compatible with most storage systems
- Sterilized by gamma irradiation
The closures and tubes are both manufactured of polypropylene having the same coefficient of expansion, which further enhances the leak-proof qualities of these vials at changing temperatures
Tubes are available in three series.
Ordering information: All Cryovials® are sold by case quantity (10× 100 per case).
Packaging: Packaged in unique tempered-proof, reseal-able, safety-lock-bags of 100 and 10 bags to a case.
6 Options Available Below
Product Details & Documents
These vials are designed for storage of biological material, human or animal cells, at temperatures as low as –190 °C.
- The cap features a long skirt for one hand aseptic technique
- A super fast thread design that allows tightening or removal with a mere 1¼" turn, and an inside thread design that will not contribute to possible contamination
- Tubes are provided with white marking area for sample identification and can be color-coded by the use of a CAPINSERT®
- Compatible with most storage systems
- Sterilized by gamma irradiation
The closures and tubes are both manufactured of polypropylene having the same coefficient of expansion, which further enhances the leak-proof qualities of these vials at changing temperatures
Tubes are available in three series.
Ordering information: All Cryovials® are sold by case quantity (10× 100 per case).
Packaging: Packaged in unique tempered-proof, reseal-able, safety-lock-bags of 100 and 10 bags to a case.
Product Family Options
Series T309
Informations sur le produit
- DésignationCapacitéBottom StyleDiameter ×HAvec bouchonAvailabilityPrice
- Series T309-22.00 mlRound bottom12.5×48 mmOuiCase of 1,000$850.75Each$850.75Supplier Number: 61800-02
- Series T309-5A5.00 mlRound bottom, self-standing12.5×78 mmOuiCase of 1,000$1,003.44Each$1,003.44Supplier Number: 61800-5A
- Series T309-3A3.00 mlRound bottom, self-standing12.5×72 mmOuiDiscontinued-Supplier Number: 61800-3A
- Series T309-2A2.00 mlRound bottom, self-standing12.5×49 mmOuiCase of 1,000$989.58Each$989.58Supplier Number: 61800-2A
- Series T309-4A4.00 mlRound bottom, self-standing12.5×78 mmOuiDiscontinued-Supplier Number: 61800-4A
Series T311
Informations sur le produit
- DésignationCapacitéBottom StyleDiameter ×HAvec bouchonAvailabilityPrice
- Series T311-11.20 mlRound bottom, self-standing12.5×43 mmOuiCase of 1,000$1,196.82Each$1,196.82Supplier Number: 61802-1A
- Series T311-32.00 mlRound bottom12.5×48 mmOuiCase of 1,000$1,028.88Each$1,028.88Supplier Number: 61802-2A
- Series T311-22.00 mlRound bottom, self-standing12.5×49 mmOuiCase of 1,000$1,206.33Each$1,206.33Supplier Number: 61802-02
- Series T311-44.00 mlRound bottom12.5×72 mmOuiDiscontinued-Supplier Number: 61802-3A