C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
Certificates
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
With a pipetor, 15 to 20 microlilter cell-containing sample is introduced into the counting chamber through one of the ports
These Disposable Cell Counters are an enclosed chamber with two ports for sample introduction. The chamber contains precisely spaced lines in a grid pattern. There are two counting chambers per slide for replicates The counting grid pattern is the Improved Neubauer, as in a common Hemacytometer. The consistent pattern design allows the standard cell counting procedure to be followed.
With a pipetor, 15 to 20 microlilter cell-containing sample is introduced into the counting chamber through one of the ports. Liquid spreads inside the chamber, replaces air, which escapes through the opposing port. The cell counting chamber is placed under a phase contrast optical microscope with a typical magnification of 100. The microscope is focused both onto the grid pattern and the cell particles. Standard protocols are followed for cell counting.
Liquid spreads inside the chamber, replaces air, which escapes through the opposing port. The cell counting chamber is placed under a phase contrast optical microscope with a typical magnification of 100. The microscope is focused both onto the grid pattern and the cell particles. Standard protocols are followed for cell counting.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Cellometer Disposable Counting Chamber, Plastic Base
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $1,461.68
    $1,461.68
    Log in for availability
    Each
    Each - $1,461.68
    $1,461.68
    Box of 100
       $1,461.68
    Each
       $1,461.68
    Supplier Number: 63561-100
  • Cellometer Disposable Counting Chamber, Plastic Base
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $477.09
    $477.09
    Log in for availability
    Each
    Each - $477.09
    $477.09
    Box of 25
       $477.09
    Each
       $477.09
    Supplier Number: 63561-25