EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences | Avantor
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Certificates
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Modules have 24 slots labeled with letters from A-X where the grids can be stored and rotated for easy access as well as 3 additional cavities for block storage .
Available with and without a unique identification number.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Dial A Grid-N-Block storage box
    Discontinued
    Supplier Number: 71158-01
  • Dial A Grid-N-Block storage box
    Discontinued
    Supplier Number: 71158-05
  • Dial A Grid-N-Block storage box
    Discontinued
    Supplier Number: 71158-10
  • Dial A Grid-N-Block storage box with unique number
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $50.46
    $50.46
    Each
       $50.46
    Supplier Number: 71158-15
  • Dial A Grid-N-Block storage box with unique number
    Discontinued
    Supplier Number: 71158-20
  • Dial A Grid-N-Block storage box with unique number
    Discontinued
    Supplier Number: 71158-25