Flat Embedding with Convenient Box, Electron Microscopy Sciences | Avantor
Flat Embedding with Convenient Box, Electron Microscopy Sciences
Flat Embedding with Convenient Box, Electron Microscopy Sciences
Certificates
Flat Embedding with Convenient Box, Electron Microscopy Sciences
Flat Embedding with Convenient Box, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
No more stickiness with new storage box. Most resins are no longer a challenge to flat embed when used in combination with the Cocoon box. The system withstands temperatures up to 70 °C, and our flat embedding of many resins is made easier.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Double End Flat Embedding Mold with Cocoon Box
    Discontinued
    Supplier Number: 70870-CB
  • 6 mm Flat Embedding Mold with Cocoon Box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $49.29
    $49.29
    Each
       $49.29
    Supplier Number: 70901-CB